2-Bromo-4-(difluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific

PC57721-1G PC57721-250MG
APOSPC57721-1GEA 719 EUR
APOSPC57721-1G APOSPC57721-250MG
2-Bromo-4-(difluoromethyl)benzoic acid 95%
2-Bromo-4-(difluoromethyl)benzoic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251.025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32070680
CAS-Nummer: 1784852-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR