2-Bromo-4-(difluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
PC57721-1G
PC57721-250MG
APOSPC57721-1GEA
719
EUR
APOSPC57721-1G
APOSPC57721-250MG
2-Bromo-4-(difluoromethyl)benzoic acid 95%
2-Bromo-4-(difluoromethyl)benzoic acid
Formel:
C₈H₅BrF₂O₂ Molecular Weight: 251.025 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32070680 CAS-Nummer: 1784852-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...