3-Bromo-4-(difluoromethyl)benzoic acid 95+%

Lieferant: Apollo Scientific
Achtung

PC57723-250MG
APOSPC57723-250MGEA 72.5 EUR
APOSPC57723-250MG
3-Bromo-4-(difluoromethyl)benzoic acid 95+%
3-Bromo-4-(difluoromethyl)benzoic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,025 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11110865
CAS-Nummer: 1131615-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR