4-Bromo-3-(difluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
PC57724-1G
PC57724-250MG
APOSPC57724-1GEA
654
EUR
APOSPC57724-1G
APOSPC57724-250MG
4-Bromo-3-(difluoromethyl)benzoic acid 95%
4-Bromo-3-(difluoromethyl)benzoic acid
Formel:
C₈H₅BrF₂O₂ Molecular Weight: 251.025 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32070682 CAS-Nummer: 2167386-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...