2-Bromo-5-(difluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC57728-250MG
PC57728-1G
PC57728-5G
APOSPC57728-250MGEA
174
EUR
APOSPC57728-250MG
APOSPC57728-1G
APOSPC57728-5G
2-Bromo-5-(difluoromethyl)benzoic acid 95%
2-Bromo-5-(difluoromethyl)benzoic acid
Formel:
C₈H₅BrF₂O₂ Molecular Weight: 251.02 g/mol |
MDL:
MFCD32070685 CAS-Nummer: 1783329-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...