2-Bromo-5-(difluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC57728-250MG PC57728-1G PC57728-5G
APOSPC57728-250MGEA 174 EUR
APOSPC57728-250MG APOSPC57728-1G APOSPC57728-5G
2-Bromo-5-(difluoromethyl)benzoic acid 95%
2-Bromo-5-(difluoromethyl)benzoic acid
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251.02 g/mol
MDL: MFCD32070685
CAS-Nummer: 1783329-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR