4-Bromo-3-(difluoromethyl)benzonitrile 95%
Lieferant: Apollo Scientific
|
Gefahr
|
PC57760-1G
APOSPC57760-1GEA
522
EUR
APOSPC57760-1G
4-Bromo-3-(difluoromethyl)benzonitrile 95%
4-Bromo-3-(difluoromethyl)benzonitrile
Formel:
C₈H₄BrF₂N Molecular Weight: 232,02 g/mol |
MDL:
MFCD18391065 CAS-Nummer: 1254034-56-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...