4-Bromo-3-(difluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC57769-1G
APOSPC57769-1GEA 609 EUR
APOSPC57769-1G
4-Bromo-3-(difluoromethyl)benzyl bromide 95%
4-Bromo-3-(difluoromethyl)benzyl bromide
Formel: C₈H₆Br₂F₂
Molecular Weight: 299,94 g/mol
MDL: MFCD18391075
CAS-Nummer: 1261580-21-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR