5-Bromo-2-(difluoromethyl)benzyl bromide ≥95%

Lieferant: Apollo Scientific

PC57771-250MG PC57771-1G
APOSPC57771-250MGEA 219 EUR
APOSPC57771-250MG APOSPC57771-1G
5-Bromo-2-(difluoromethyl)benzyl bromide ≥95%
5-Bromo-2-(difluoromethyl)benzyl bromide
Formel: C₈H₆Br₂F₂
MDL: MFCD18391076
CAS-Nummer: 1261760-28-4
UN: 3265

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR