2-Bromo-5-(difluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC57773-250MG PC57773-1G PC57773-5G
APOSPC57773-250MGEA 192 EUR
APOSPC57773-250MG APOSPC57773-1G APOSPC57773-5G
2-Bromo-5-(difluoromethyl)benzyl bromide 95%
2-Bromo-5-(difluoromethyl)benzyl bromide
Formel: C₈H₆Br₂F₂
MDL: MFCD18391069
CAS-Nummer: 1261441-93-3
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR