2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
Lieferant: Apollo Scientific
|
Achtung
|
PC5778-1G
PC5778-5G
APOSPC5778-1GEA
157
EUR
APOSPC5778-1G
APOSPC5778-5G
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
2-Chloro-1,1,1,6,6,6-hexafluorohexa-2,4-diene
Formel:
C₆H₃ClF₆ Molecular Weight: 224.531 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08461616 CAS-Nummer: 885276-10-8 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...