5-(Trifluoromethyl)benzothiophene-2-carboxamide

Lieferant: Apollo Scientific
Achtung

PC57787-1G PC57787-250MG
APOSPC57787-1GEA 132 EUR
APOSPC57787-1G APOSPC57787-250MG
5-(Trifluoromethyl)benzothiophene-2-carboxamide
5-(Trifluoromethyl)benzothiophene-2-carboxamide
Formel: C₁₀H₆F₃NOS
Lagertemperatur: Raumtemperatur
MDL: MFCD31567862
CAS-Nummer: 1609382-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR