4-(Trifluoromethyl)benzothiophene-2-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
PC57791-100MG
PC57791-1G
PC57791-250MG
APOSPC57791-100MGEA
21.9
EUR
APOSPC57791-100MG
APOSPC57791-1G
APOSPC57791-250MG
4-(Trifluoromethyl)benzothiophene-2-carbonitrile
4-(Trifluoromethyl)benzothiophene-2-carbonitrile
Formel:
C₁₀H₄F₃NS Schmelzpunkt: 85…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD31567871 CAS-Nummer: 2115333-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...