4-(Trifluoromethyl)benzothiophene-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC57793-1G PC57793-250MG
APOSPC57793-1GEA 0 EUR
APOSPC57793-1G APOSPC57793-250MG
4-(Trifluoromethyl)benzothiophene-2-carbaldehyde
4-(Trifluoromethyl)benzothiophene-2-carbaldehyde
Formel: C₁₀H₅F₃OS
Lagertemperatur: Raumtemperatur
MDL: MFCD27932089
CAS-Nummer: 908566-20-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR