2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene

Lieferant: Apollo Scientific
Gefahr

PC57794-100MG PC57794-1G PC57794-250MG
APOSPC57794-100MGEA 31.9 EUR
APOSPC57794-100MG APOSPC57794-1G APOSPC57794-250MG
2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene
2-(Bromomethyl)-4-(trifluoromethyl)benzothiophene
Formel: C₁₀H₆BrF₃S
Molecular Weight: 295.119 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31567869
CAS-Nummer: 2377846-69-8
UN: 3261
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR