(±)-2-Fluor-α-methylbenzylamin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Fluorophenyl)ethylamine
PC5780-1G
APOSPC5780-1GEA
21.9
EUR
APOSPC5780-1G
(±)-2-Fluor-α-methylbenzylamin 97%
(±)-2-Fluor-α-methylbenzylamin
Formel:
C₈H₁₀FN Molecular Weight: 139,17 g/mol Siedepunkt: 70,4…73.5 °C Flammpunkt: 70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038307 CAS-Nummer: 74788-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...