3-Chlor-5-(trifluormethoxy)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC5790-1G PC5790-5G
APOSPC5790-1GEA 0 EUR
APOSPC5790-1G APOSPC5790-5G
3-Chlor-5-(trifluormethoxy)benzylbromid 97%
3-Chlor-5-(trifluormethoxy)benzylbromid
Formel: C₈H₅BrClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD06660311
CAS-Nummer: 886503-33-9
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR