1H-Perfluorohexane-1,1-diol
Lieferant: Apollo Scientific
|
Achtung
|
PC5791-1G
APOSPC5791-1GEA
0
EUR
APOSPC5791-1G
1H-Perfluorohexane-1,1-diol
1H-Perfluorohexane-1,1-diol
Formel:
C₆H₃F₁₁O₂ Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00437976 CAS-Nummer: 904037-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...