2-Fluor-6-pyridon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoro-6-hydroxypyridine, 6-Fluoropyridin-2-ol

PC5794-1G
APOSPC5794-1GEA 21.9 EUR
APOSPC5794-1G
2-Fluor-6-pyridon 97%
2-Fluor-6-pyridon
Formel: C₅H₄FNO
Molecular Weight: 113,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03092938
CAS-Nummer: 50543-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR