3-Cyan-4-fluoranilin 97%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril

PC5823-25G PC5823-100G
APOSPC5823-25GEA 125 EUR
APOSPC5823-25G APOSPC5823-100G
3-Cyan-4-fluoranilin 97%
3-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
Schmelzpunkt: 88…96 °C
MDL: MFCD00277872
CAS-Nummer: 53312-81-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR