1,2-Dichloro-4-iodoperfluorobutane
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Dichloro-1,1,2,3,3,4,4-heptafluoro-4-iodobutane
PC5827-5G
APOSPC5827-5GEA
170
EUR
APOSPC5827-5G
1,2-Dichloro-4-iodoperfluorobutane
1,2-Dichloro-4-iodoperfluorobutane
Formel:
C₄Cl₂F₇I Molecular Weight: 378,842 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042241 CAS-Nummer: 678-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...