2-Bromo-6-(trifluoromethyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC5850-1G
APOSPC5850-1GEA 0 EUR
APOSPC5850-1G
2-Bromo-6-(trifluoromethyl)benzyl bromide
2-Bromo-6-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Lagertemperatur: Kühlschrank
MDL: MFCD12026460
CAS-Nummer: 69902-84-7
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR