5-Cyan-2-fluor-6-picolin
Lieferant: Apollo Scientific
|
Achtung
|
PC5881-1G
APOSPC5881-1GEA
91.5
EUR
APOSPC5881-1G
5-Cyan-2-fluor-6-picolin
5-Cyan-2-fluor-6-picolin
Formel:
C₇H₅FN₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD03095096 CAS-Nummer: 375368-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...