2,3,5,6-Tetrafluoro-1,4-phenylenediamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Diamino-2,3,5,6-tetrafluorobenzene

PC5882-1G
APOSPC5882-1GEA 388 EUR
APOSPC5882-1G
2,3,5,6-Tetrafluoro-1,4-phenylenediamine
2,3,5,6-Tetrafluoro-1,4-phenylenediamine
Formel: C₆H₄F₄N₂
Molecular Weight: 180,11 g/mol
Schmelzpunkt: 142…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039767
CAS-Nummer: 1198-64-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR