1-(2-Fluorophenyl)propylamine 97%

Lieferant: Apollo Scientific

Synonyme: 1-(2-Fluorophenyl)propan-1-amine

PC5883-1G
APOSPC5883-1GEA 347 EUR
APOSPC5883-1G
1-(2-Fluorophenyl)propylamine 97%
1-(2-Fluorophenyl)propylamine
Formel: C₉H₁₂FN
Molecular Weight: 153.197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784283
CAS-Nummer: 473249-01-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR