3-Fluor-5-iodanilin

Lieferant: Apollo Scientific
Achtung

PC5888-1G
APOSPC5888-1GEA 147 EUR
APOSPC5888-1G
3-Fluor-5-iodanilin
3-Fluor-5-iodanilin
Formel: C₆H₅FIN
Flammpunkt: >110 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04116466
CAS-Nummer: 660-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR