1-[2-(Trifluoromethyl)quinol-4-yl]homopiperazine 97%

Lieferant: Apollo Scientific
Achtung

PC5893-500MG
APOSPC5893-500MGEA 279 EUR
APOSPC5893-500MG
1-[2-(Trifluoromethyl)quinol-4-yl]homopiperazine 97%
1-[2-(Trifluoromethyl)quinol-4-yl]homopiperazine
Formel: C₁₅H₁₆F₃N₃
Molecular Weight: 295,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094309
CAS-Nummer: 401566-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR