2-Fluor-5-(trifluormethoxy)anilin 99%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5900-1G
PC5900-25G
PC5900-5G
APOSPC5900-1GEA
0
EUR
APOSPC5900-1G
APOSPC5900-25G
APOSPC5900-5G
2-Fluor-5-(trifluormethoxy)anilin 99%
2-Fluor-5-(trifluormethoxy)anilin
Formel:
C₇H₅F₄NO Siedepunkt: 64…65 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832254 CAS-Nummer: 116369-23-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...