3,5-Difluortoluol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,3-Difluor-5-methylbenzol

PC5913-5G
APOSPC5913-5GEA 21.9 EUR
APOSPC5913-5G
3,5-Difluortoluol 98%
3,5-Difluortoluol
Formel: C₇H₆F₂
Molecular Weight: 128,12 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00797323
CAS-Nummer: 117358-51-7
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR