5-Cyano-2-fluorobenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC5926-5G
APOSPC5926-5GEA
21.9
EUR
APOSPC5926-5G
5-Cyano-2-fluorobenzoic acid
5-Cyano-2-fluorobenzoic acid
Formel:
C₈H₄FNO₂ Schmelzpunkt: 187…189 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03094321 CAS-Nummer: 146328-87-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...