4-(Difluoromethoxy)phenylacetylene 95%

Lieferant: Apollo Scientific
Achtung

PC5954-1G PC5954-250MG
APOSPC5954-1GEA 638 EUR
APOSPC5954-1G APOSPC5954-250MG
4-(Difluoromethoxy)phenylacetylene 95%
4-(Difluoromethoxy)phenylacetylene
Formel: C₉H₆F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03094335
CAS-Nummer: 519059-04-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR