Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Dimethyl (3,3-difluoro-2-oxoheptyl)phosphonate, (3,3-Difluoro-2-oxoheptyl)phosphonic acid Dimethyl ester

PC5961-5G
APOSPC5961-5GEA 157 EUR
APOSPC5961-5G
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate
Formel: C₉H₁₇F₂O₄P
Molecular Weight: 258.20 g/mol
Siedepunkt: 108…110 °C/0.4mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD11707108
CAS-Nummer: 50889-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR