2,5-Bis(trifluormethyl)benzylalkohol 98%
Lieferant: Apollo Scientific
PC5967-25G
PC5967-5G
APOSPC5967-25GEA
310
EUR
APOSPC5967-25G
APOSPC5967-5G
2,5-Bis(trifluormethyl)benzylalkohol 98%
2,5-Bis(trifluormethyl)benzylalkohol
Formel:
C₉H₆F₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00799484 CAS-Nummer: 302911-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...