2,5-Bis(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific

PC5967-25G PC5967-5G
APOSPC5967-25GEA 310 EUR
APOSPC5967-25G APOSPC5967-5G
2,5-Bis(trifluormethyl)benzylalkohol 98%
2,5-Bis(trifluormethyl)benzylalkohol
Formel: C₉H₆F₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00799484
CAS-Nummer: 302911-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR