1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododec-1-yl 2-methylprop-2-enoate, 2-(Perfluorodec-1-yl)ethyl methacrylate
PC5969P-5G
APOSPC5969P-5GEA
270
EUR
APOSPC5969P-5G
1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate 97%
1H,1H,2H,2H-Henicosafluorododec-1-yl methacrylate
Formel:
C₁₆H₉F₂₁O₂ Molecular Weight: 632.208 g/mol Siedepunkt: 95…100 °C (0.1 mm Hg) Schmelzpunkt: 40…45 °C Flammpunkt: 113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00236095 CAS-Nummer: 2144-54-9 EINECS: 218-408-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...