2,5-Bis(trifluormethyl)phenylacetonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC5971-1G
APOSPC5971-1GEA
21.9
EUR
APOSPC5971-1G
2,5-Bis(trifluormethyl)phenylacetonitril 98%
2,5-Bis(trifluormethyl)phenylacetonitril
Formel:
C₁₀H₅F₆N Lagertemperatur: Raumtemperatur |
CAS-Nummer:
302911-99-5 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...