4-Fluoro-1H-indole-7-carbonitrile 97%
Lieferant: Apollo Scientific
Synonyme:
7-Cyano-4-fluoro-1H-indole
PC5980-250MG
APOSPC5980-250MGEA
77
EUR
APOSPC5980-250MG
4-Fluoro-1H-indole-7-carbonitrile 97%
4-Fluoro-1H-indole-7-carbonitrile
Formel:
C₉H₅FN₂ Molecular Weight: 160,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845597 CAS-Nummer: 313337-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...