Perfluoro(2-ethoxyethane)sulphonic acid 97%

Lieferant: Apollo Scientific
Gefahr

PC6005-25G
APOSPC6005-25GEA 575 EUR
APOSPC6005-25G
Perfluoro(2-ethoxyethane)sulphonic acid 97%
Perfluoro(2-ethoxyethane)sulphonic acid
Formel: C₄F₉HO₄S
Siedepunkt: 198 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00153237
CAS-Nummer: 113507-82-7
UN: 2586
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR