4-Brom-2-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

PC6013-1G PC6013-5G PC6013-25G PC6013-10G
APOSPC6013-1GEA 21.9 EUR
APOSPC6013-1G APOSPC6013-5G APOSPC6013-25G APOSPC6013-10G
4-Brom-2-(trifluormethyl)benzonitril 98%
4-Brom-2-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD06659466
CAS-Nummer: 191165-13-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR