4-Bromo-3-(trifluoromethyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-5-(bromomethyl)benzotrifluoride, 1-Bromo-4-(bromomethyl)-2-(trifluoromethyl)benzene
PC6025-5G
PC6025-1G
PC6025-250MG
APOSPC6025-5GEA
405
EUR
APOSPC6025-5G
APOSPC6025-1G
APOSPC6025-250MG
4-Bromo-3-(trifluoromethyl)benzyl bromide
4-Bromo-3-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅Br₂F₃ Schmelzpunkt: 52…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059128 CAS-Nummer: 1159512-68-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...