4-Bromo-3-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific
Achtung

PC6026-1G PC6026-5G
APOSPC6026-1GEA 0 EUR
APOSPC6026-1G APOSPC6026-5G
4-Bromo-3-(trifluoromethyl)phenylacetonitrile
4-Bromo-3-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD12026483
CAS-Nummer: 1159512-69-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR