4-Bromo-3-(trifluoromethyl)benzylamine 98%

Lieferant: Apollo Scientific
Gefahr

PC6027-1G PC6027-250MG PC6027-5G
APOSPC6027-1GEA 0 EUR
APOSPC6027-1G APOSPC6027-250MG APOSPC6027-5G
4-Bromo-3-(trifluoromethyl)benzylamine 98%
Formel: C₈H₇BrF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD12026484
CAS-Nummer: 1159512-70-5
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR