1-(2,3-Dichlorophenyl)-3-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl]guanidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2,3-Dichlorophenyl)-N'-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl]guanidine, 2-[N'-(2,3-Dichlorophenyl)carbamimidamido]-6-(trifluoromethyl)pyrimidin-4-ol
PC6032-5G
APOSPC6032-5GEA
192
EUR
APOSPC6032-5G
1-(2,3-Dichlorophenyl)-3-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl]guanidine
1-(2,3-Dichlorophenyl)-3-[4-hydroxy-6-(trifluoromethyl)pyrimidin-2-yl]guanidine
Formel:
C₁₂H₈Cl₂F₃N₅O Molecular Weight: 366,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05150372 CAS-Nummer: 618092-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...