N-Ethyl-2-(trifluormethoxy)anilin 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoromethoxy)ethylaminobenzene

PC6038-250MG
APOSPC6038-250MGEA 21.9 EUR
APOSPC6038-250MG
N-Ethyl-2-(trifluormethoxy)anilin 95+%
N-Ethyl-2-(trifluormethoxy)anilin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Siedepunkt: 42…44 °C/2,0mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00204184
CAS-Nummer: 175278-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR