6-Fluor-3-(4-piperidinyl)-1,2-benzoxazolhydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: Paliperidone Related Compound B

PC6042-1G
APOSPC6042-1GEA 21.9 EUR
APOSPC6042-1G
6-Fluor-3-(4-piperidinyl)-1,2-benzoxazolhydrochlorid
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole hydrochloride
Formel: C₁₂H₁₄ClFN₂O
Molecular Weight: 256,7000 g/mol
Schmelzpunkt: 293…295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06658529
CAS-Nummer: 84163-13-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR