2-(Trifluoromethyl)thiazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

PC6043-1G
APOSPC6043-1GEA 0 EUR
APOSPC6043-1G
2-(Trifluoromethyl)thiazole-4-carboxylic acid
2-(Trifluoromethyl)thiazole-4-carboxylic acid
Formel: C₅H₂F₃NO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD09258845
CAS-Nummer: 915030-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR