N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Boc-3-fluoro-L-phenylalanine, Boc-Phe(3-F)-OH
PC6045-1G
APOSPC6045-1GEA
21.9
EUR
APOSPC6045-1G
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine
Formel:
C₁₄H₁₈FNO₄ Schmelzpunkt: 75…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00672522 CAS-Nummer: 114873-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...