4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid

Lieferant: Apollo Scientific
Achtung

PC6055-1G PC6055-5G
APOSPC6055-1GEA 0 EUR
APOSPC6055-1G APOSPC6055-5G
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formel: C₁₃H₈F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00955325
CAS-Nummer: 773108-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR