2-Fluoro-6-nitrophenyl methyl sulphone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-2-(methylsulphonyl)nitrobenzene, 1-Fluoro-2-(methylsulphonyl)-3-nitrobenzene
PC6067-250MG
APOSPC6067-250MGEA
198
EUR
APOSPC6067-250MG
2-Fluoro-6-nitrophenyl methyl sulphone
2-Fluoro-6-nitrophenyl methyl sulphone
Formel:
C₇H₆FNO₄S Molecular Weight: 219,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12922837 CAS-Nummer: 1210693-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...