3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluormethyl)anilin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-[3-Amino-5-(trifluoromethyl)phenyl]-4-methylimidazole
PC6078-25G
APOSPC6078-25GEA
63.8
EUR
APOSPC6078-25G
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluormethyl)anilin 98%
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline
Formel:
C11H10F3N3 Molecular Weight: 241,2124 g/mol Schmelzpunkt: 130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846236 CAS-Nummer: 641571-11-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...